Tripropargylamine 97%
Dodavatel: Thermo Scientific
|
Varování
|
H53459.06
H53459.14
H53459.03
H53459.06EA
6590
CZK
H53459.06
H53459.14
H53459.03
Tripropargylamine 97%
Tripropargylamine
Purity: 97%
CAS No.: 6921-29-5
Molecular Formula: C9H9N
Formula Weight: 131.18
Flash Point: 57°(135°F)
Refractive Index: 1.484
Boiling Point: 79-85°/11mm
UN No.: UN1993
MDL No.: MFCD00008577
Vzorec:
C₉H₉N Bod varu: 79…85 °C (11 mmHg) Hustota: 0,927 Bod vzplanutí: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008577 CAS číslo: 6921-29-5 EINECS: 230-038-5 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...