Tisk…

Propargyl benzoate 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Prop-2-yn-1-yl benzoate

H53444.06 H53444.14
H53444.06EA 1660 CZK
H53444.06 H53444.14
Propargyl benzoate 98%
Propargyl benzoate

Purity: 98% CAS No.: 6750-04-5 Molecular Formula: C10H8O2 Formula Weight: 160.17 Refractive Index: 1.5320 Boiling Point: 225-226° MDL No.: MFCD00996341

Vzorec: C₁₀H₈O₂
MW: 160.17 g/mol
Bod varu: 225…226 °C
Hustota: 1.106
Storage Temperature: Ambient
MDL Number: MFCD00996341
CAS číslo: 6750-04-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR