Tisk…

tert-Butyl pivalate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: tert-Butyl trimethylacetate

H53395.06 H53395.14 H53395.22
H53395.06EA 1140 CZK
H53395.06 H53395.14 H53395.22
tert-Butyl pivalate 98%
tert-Butyl pivalate

Purity: 98% CAS No.: 16474-43-4 Molecular Formula: C9H18O2 Formula Weight: 158.24 Flash Point: 62°(144°F) Refractive Index: 1.392 Boiling Point: 134-135° MDL No.: MFCD01861974

Vzorec: C₉H₁₈O₂
MW: 158,24 g/mol
Bod varu: 134…135 °C
Hustota: 0,813
Bod vzplanutí: >62 °C (144 °F)
Storage Temperature: Ambient
MDL Number: MFCD01861974
CAS číslo: 16474-43-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR