1-Methylsulphonyl-1H-benzotriazole 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-(Methylsulphonyl)-1H-benzo[d][1,2,3]triazole
H53386.03
H53386.06
H53386.14
H53386.03EA
1310
CZK
H53386.03
H53386.06
H53386.14
1-Methylsulphonyl-1H-benzotriazole 97%
1-Methylsulphonyl-1H-benzotriazole
Purity: 97%
CAS No.: 37073-15-7
Molecular Formula: C7H7N3O2S
Formula Weight: 197.21
Melting Point: 106-108° dec.
MDL No.: MFCD00595638
Vzorec:
C₇H₇N₃O₂S MW: 197,21 g/mol Bod tání: 106…108 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00595638 CAS číslo: 37073-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...