Boc-Arg-OH 98%
Dodavatel: Thermo Scientific
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-5-guanidinopentanoic acid
H53379.03
H53379.06
H53379.03EA
1780
CZK
H53379.03
H53379.06
Boc-Arg-OH 98%
Boc-Arg-OH
Purity: 98%
CAS No.: 13726-76-6
Molecular Formula: C11H22N4O4
Formula Weight: 274.32
Flash Point: 113°(235°F)
Beilstein Reference: 2419628
Optical Rotation: -6.5° (c=1 in acetic acid)
MDL No.: MFCD00042632
Vzorec:
C₁₁H₂₂N₄O₄ MW: 274,321 g/mol Bod vzplanutí: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042632 CAS číslo: 13726-76-6 EINECS: 237-295-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...