(S)-1-(Boc-amino)-2-propanol 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
tert-Butyl (S)-(2-hydroxypropyl)carbamate
H52826.06
H52826.14
H52826.06EA
2020
CZK
H52826.06
H52826.14
(S)-1-(Boc-amino)-2-propanol 97%
(S)-1-(Boc-amino)-2-propanol
Purity: 97%
CAS No.: 167938-56-9
Molecular Formula: C8H17NO3
Formula Weight: 175.23
MDL No.: MFCD04974340
Vzorec:
C₈H₁₇NO₃ MW: 175,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04974340 CAS číslo: 167938-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...