Tisk…

Boc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid 97%

Dodavatel: Thermo Scientific
Varování

H52806.03 H52806.MD
H52806.03EA 26360 CZK
H52806.03 H52806.MD
Boc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid 97%
Boc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid

Purity: 97% CAS No.: 73397-28-1 Molecular Formula: C12H23NO5 Formula Weight: 261.32 MDL No.: MFCD04972275

Vzorec: C₁₂H₂₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD04972275
CAS číslo: 73397-28-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR