Tisk…

tert-Butyl 2-phenylpiperazine-1-carboxylate 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Phenylpiperazine, N1-BOC protected, 1-Boc-2-phenylpiperazine, 1-(tert-Butoxycarbonyl)-2-phenylpiperazine, 2-Methyl-2-propanyl 2-phenyl-1-piperazinecarboxylate

H52797.MD H52797.03
H52797.MDEA 3750 CZK
H52797.MD H52797.03
tert-Butyl 2-phenylpiperazine-1-carboxylate 97%
tert-Butyl 2-phenylpiperazine-1-carboxylate

Purity: 97% CAS No.: 886766-60-5 Molecular Formula: C15H22N2O2 Formula Weight: 262.35 MDL No.: MFCD07367734

Vzorec: C₁₅H₂₂N₂O₂
MW: 262,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367734
CAS číslo: 886766-60-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR