Tisk…

1-Boc-azetidine-3-carboxaldehyde 97%

Dodavatel: Thermo Fisher Scientific
Varování

H52794.03 H52794.06
H52794.03EA 5940 CZK
H52794.03 H52794.06
1-Boc-azetidine-3-carboxaldehyde 97%
1-Boc-azetidine-3-carboxaldehyde

Purity: 97% CAS No.: 177947-96-5 Molecular Formula: C9H15NO3 Formula Weight: 185.22 Storage Temperature: 2°C to 8°C Boiling Point: 77-82°/2mm MDL No.: MFCD06656142

Vzorec: C₉H₁₅NO₃
Bod varu: 77…82 °C (2 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD06656142
CAS číslo: 177947-96-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR