Boc-(S)-thiazolidine-2-carboxylic acid 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(S)-3-(tert-Butoxycarbonyl)thiazolidine-2-carboxylic acid
H52793.03
H52793.06
H52793.03EA
4160
CZK
H52793.03
H52793.06
Boc-(S)-thiazolidine-2-carboxylic acid 97%
Boc-(S)-thiazolidine-2-carboxylic acid
Purity: 97%
CAS No.: 891192-95-3
Molecular Formula: C9H15NO4S
Formula Weight: 233.28
Melting Point: 90-93°
Optical Rotation: -47° (c=5 in methanol)
MDL No.: MFCD02682342
Vzorec:
C₉H₁₅NO₄S MW: 233.29 g/mol Bod tání: 90…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD02682342 CAS číslo: 891192-95-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...