tert-Butyl (piperidin-4-ylmethyl)carbamate 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(4-Piperidylmethyl)carbamic acid tert-Butyl ester, tert-Butyl (4-Piperidylmethyl)carbamate, 4-(tert-Butoxycarbonylaminomethyl)piperidine, 4-(Boc-aminomethyl)piperidine
H52725.03
H52725.06
H52725.03EA
2660
CZK
H52725.03
H52725.06
tert-Butyl (piperidin-4-ylmethyl)carbamate 97%
tert-Butyl (piperidin-4-ylmethyl)carbamate
Purity: 97%
CAS No.: 135632-53-0
Molecular Formula: C11H22N2O2
Formula Weight: 214.31
Melting Point: 106-108°
UN No.: UN3259
MDL No.: MFCD01631214
Vzorec:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol Bod tání: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631214 CAS číslo: 135632-53-0 UN: 3259 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...