Tisk…

Boc-(2S,3S)-3-amino-2-hydroxy-5-methylhexanoic acid 97%

Dodavatel: Thermo Scientific
Varování

H52569.MD H52569.03
H52569.MDEA 6590 CZK
H52569.MD H52569.03
Boc-(2S,3S)-3-amino-2-hydroxy-5-methylhexanoic acid 97%
Boc-(2S,3S)-3-amino-2-hydroxy-5-methylhexanoic acid

Purity: 97% CAS No.: 73397-27-0 Molecular Formula: C12H23NO5 Formula Weight: 261.32 MDL No.: MFCD04974450

Vzorec: C₁₂H₂₃NO₅
MW: 261,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974450
CAS číslo: 73397-27-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR