(S)-2-Amino-3-methoxypropanoic acid hydrochloride 97%
Dodavatel: Thermo Fisher Scientific
H52562.MD
H52562.03
H52562.MDEA
4290
CZK
H52562.MD
H52562.03
(S)-2-Amino-3-methoxypropanoic acid hydrochloride 97%
(S)-2-Amino-3-methoxypropanoic acid hydrochloride
Purity: 97%
CAS No.: 336100-47-1
Molecular Formula: C4H9NO3.HCl
Formula Weight: 155.58
Melting Point: 182-183°
MDL No.: MFCD03840366
Vzorec:
C₄H₁₀ClNO₃ Bod tání: 182…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840366 CAS číslo: 336100-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...