(R)-1,4-Diazabicyclo[4.3.0]nonane 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H52554.MD
H52554.03
H52554.MDEA
4120
CZK
H52554.MD
H52554.03
(R)-1,4-Diazabicyclo[4.3.0]nonane 97%
(R)-1,4-Diazabicyclo[4.3.0]nonane
Purity: 97%
CAS No.: 96193-27-0
Molecular Formula: C7H14N2
Formula Weight: 126.20
Boiling Point: 65-69°/3mm
UN No.: UN3267
MDL No.: MFCD09752820
Vzorec:
C₇H₁₄N₂ MW: 126,20 g/mol Bod varu: 65…69 °C (3 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD09752820 CAS číslo: 96193-27-0 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...