Tisk…

4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Fluoro-3-(hydroxymethyl)phenylboronic acid, 4-Fluoro-3-(hydroxymethyl)benzeneboronic acid

H52487.MD H52487.03
H52487.MDEA 2680 CZK
H52487.MD H52487.03
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid 98%
4-Fluoro-3-(hydroxymethyl)benzeneboronic acid

Purity: 98% CAS No.: 481681-02-1 Molecular Formula: C7H8BFO3 Formula Weight: 169.95 Melting Point: 124-126° MDL No.: MFCD08235075

Vzorec: C₇H₈BFO₃
MW: 169,95 g/mol
Bod tání: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD08235075
CAS číslo: 481681-02-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR