Tisk…

3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: (3-(((tert-Butoxycarbonyl)amino)methyl)phenyl)boronic acid, 3-(Aminomethyl)benzeneboronic acid, N-BOC protected

H52453.MD H52453.03
H52453.MDEA 1510 CZK
H52453.MD H52453.03
3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid 97%
3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid

Purity: 97% CAS No.: 199609-62-6 Molecular Formula: C12H18BNO4 Formula Weight: 251.09 Melting Point: 188-190° MDL No.: MFCD06246052

Vzorec: C₁₂H₁₈BNO₄
MW: 251,09 g/mol
Bod tání: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD06246052
CAS číslo: 199609-62-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR