(R)-3-Amino-1-benzylpiperidine 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H52425.03
H52425.06
H52425.03EA
2780
CZK
H52425.03
H52425.06
(R)-3-Amino-1-benzylpiperidine 97%
(R)-3-Amino-1-benzylpiperidine
Purity: 97%
CAS No.: 168466-84-0
Molecular Formula: C12H18N2
Formula Weight: 190.29
MDL No.: MFCD03093374
Vzorec:
C₁₂H₁₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD03093374 CAS číslo: 168466-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...