Tisk…

(S)-1-Boc-2-isopropylpiperazine 97%

Dodavatel: Thermo Fisher Scientific
Varování

H52424.MD H52424.03
H52424.MDEA 4770 CZK
H52424.MD H52424.03
(S)-1-Boc-2-isopropylpiperazine 97%
(S)-1-Boc-2-isopropylpiperazine

Purity: 97% CAS No.: 674792-05-3 Molecular Formula: C12H24N2O2 Formula Weight: 228.33 MDL No.: MFCD03787918

Vzorec: C₁₂H₂₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03787918
CAS číslo: 674792-05-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR