Tisk…

Boc-aza-DL-leucine 97%

Dodavatel: Thermo Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)-3-(dimethylamino)propanoic acid

H52412.03 H52412.06
H52412.03EA 3430 CZK
H52412.03 H52412.06
Boc-aza-DL-leucine 97%
Boc-aza-DL-leucine

Purity: 97% CAS No.: 851653-36-6 Molecular Formula: C10H20N2O4 Formula Weight: 232.28 MDL No.: MFCD04972270

Vzorec: C₁₀H₂₀N₂O₄
MW: 232,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972270
CAS číslo: 851653-36-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR