Tisk…

4-Methoxyindole-2-carboxylic acid ≥97%

Dodavatel: Thermo Scientific

H52392.MD H52392.03 H52392.06
H52392.MDEA 1630 CZK
H52392.MD H52392.03 H52392.06
4-Methoxyindole-2-carboxylic acid ≥97%
4-Methoxyindole-2-carboxylic acid

Purity: 97+% CAS No.: 103260-65-7 Molecular Formula: C10H9NO3 Formula Weight: 191.19 MDL No.: MFCD02664458

Vzorec: C₁₀H₉NO₃
MW: 191,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664458
CAS číslo: 103260-65-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR