4-(1-Methyl-4-piperidinylmethoxy)aniline 96%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H52351.MD
H52351.03
H52351.MDEA
5260
CZK
H52351.MD
H52351.03
4-(1-Methyl-4-piperidinylmethoxy)aniline 96%
4-(1-Methyl-4-piperidinylmethoxy)aniline
Purity: 96%
CAS No.: 902454-26-6
Molecular Formula: C13H20N2O
Formula Weight: 220.31
Storage Temperature: 2°C to 8°C
UN No.: UN2811
MDL No.: MFCD09909628
Vzorec:
C₁₃H₂₀N₂O MW: 220.31 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09909628 CAS číslo: 902454-26-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...