Tisk…

N-(2-Hydroxyethyl)formamide 97%

Dodavatel: Thermo Scientific
Varování

H52345.MD H52345.03 H52345.06
H52345.MDEA 3570 CZK
H52345.MD H52345.03 H52345.06
N-(2-Hydroxyethyl)formamide 97%
N-(2-Hydroxyethyl)formamide

Purity: 97% CAS No.: 693-06-1 Molecular Formula: C3H7NO2 Formula Weight: 89.09 MDL No.: MFCD00021040

Vzorec: C₃H₇NO₂
MW: 89,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021040
CAS číslo: 693-06-1
EINECS: 211-741-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR