Tisk…

5-Carbamoylpyridine-3-boronic acid pinacol ester 96%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinamide

H52330.MD H52330.03
H52330.MDEA 5700 CZK
H52330.MD H52330.03
5-Carbamoylpyridine-3-boronic acid pinacol ester 96%
5-Carbamoylpyridine-3-boronic acid pinacol ester

Purity: 96% CAS No.: 1169402-51-0 Molecular Formula: C12H17BN2O3 Formula Weight: 248.09 MDL No.: MFCD11878274

Vzorec: C₁₂H₁₇BN₂O₃
MW: 248,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11878274
CAS číslo: 1169402-51-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR