N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine ≥98%
Dodavatel: Thermo Scientific
H52309.03
H52309.06
H52309.03EA
4840
CZK
H52309.03
H52309.06
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine ≥98%
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine
Purity: 98+%
CAS No.: 51549-39-4
Molecular Formula: C23H31N5O4Si
Formula Weight: 469.62
MDL No.: MFCD04972281
Vzorec:
C₂₃H₃₁N₅O₄Si MW: 469,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04972281 CAS číslo: 51549-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...