Fmoc-D-4-pyridylalanine 95%
Dodavatel: Thermo Scientific
H52193.MD
H52193.03
H52193.MDEA
2540
CZK
H52193.MD
H52193.03
Fmoc-D-4-pyridylalanine 95%
Fmoc-D-4-pyridylalanine
Purity: 95%
CAS No.: 205528-30-9
Molecular Formula: C23H20N2O4
Formula Weight: 388.42
MDL No.: MFCD00672567
Vzorec:
C₂₃H₂₀N₂O₄ MW: 388,43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00672567 CAS číslo: 205528-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...