Tisk…

Fmoc-L-Nw-boc-beta-homolysine 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: Fmoc-β-HomoLys(Boc)-OH

H52189.MD H52189.03
H52189.MDEA 4470 CZK
H52189.MD H52189.03
Fmoc-L-Nw-boc-beta-homolysine 95%
Fmoc-L-Nw-boc-beta-homolysine

Purity: 95% CAS No.: 203854-47-1 Molecular Formula: C27H34N2O6 Formula Weight: 482.58 Beilstein Reference: 8022293 MDL No.: MFCD01863054

Vzorec: C₂₇H₃₄N₂O₆
MW: 482.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03002713
CAS číslo: 203854-47-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR