Tisk…

2-Benzyl-N-boc-L-proline 95%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (R)-2-Benzyl-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid

H52184.MD H52184.03
H52184.MDEA 2760 CZK
H52184.MD H52184.03
2-Benzyl-N-boc-L-proline 95%
2-Benzyl-N-boc-L-proline

Purity: 95% CAS No.: 706806-60-2 Molecular Formula: C17H23NO4 Formula Weight: 305.37 Melting Point: 154-156° MDL No.: MFCD06795522

Vzorec: C₁₇H₂₃NO₄
MW: 305,37 g/mol
Bod tání: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD06795522
CAS číslo: 706806-60-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR