N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine 95%
Dodavatel: Thermo Fisher Scientific
H52181.MD
H52181.03
H52181.06
H52181.MDEA
1150
CZK
H52181.MD
H52181.03
H52181.06
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine 95%
N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine
Purity: 95%
CAS No.: 57292-44-1
Molecular Formula: C14H18ClNO4
Formula Weight: 299.75
Melting Point: 110°
Beilstein Reference: 5381988
Optical Rotation: -26° (c=1 in ethyl acetate)
MDL No.: MFCD00076978
Vzorec:
C₁₄H₁₈ClNO₄ Bod tání: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00076978 CAS číslo: 57292-44-1 |