Tisk…

Fmoc-3-(2-naphthyl)-L-alanine 95%

Dodavatel: Thermo Scientific

Synonyms: Fmoc-2-Nal-OH

H52179.MD H52179.03 H52179.06
H52179.MDEA 1900 CZK
H52179.MD H52179.03 H52179.06
Fmoc-3-(2-naphthyl)-L-alanine 95%
Fmoc-3-(2-naphthyl)-L-alanine

Purity: 95% CAS No.: 112883-43-9 Molecular Formula: C28H23NO4 Formula Weight: 437.49 Beilstein Reference: 7668973 MDL No.: MFCD00144886

Vzorec: C₂₈H₂₃NO₄
MW: 437,495 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144886
CAS číslo: 112883-43-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR