Tisk…

Fmoc-L-allylglycine 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: Fmoc-Gly(allyl)-OH

H52177.03 H52177.06
H52177.03EA 5940 CZK
H52177.03 H52177.06
Fmoc-L-allylglycine 95%
Fmoc-L-allylglycine

Purity: 95% CAS No.: 146549-21-5 Molecular Formula: C20H19NO4 Formula Weight: 337.37 Physical Form: Crystalline solid Appearance: White Beilstein Reference: 5884047 MDL No.: MFCD01311749

Vzorec: C₂₀H₁₉NO₄
MW: 337.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311749
CAS číslo: 146549-21-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR