Tisk…

Fmoc-L-3,3-diphenylalanine 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3,3-diphenylpropanoic acid, Fmoc-β,β-diphenyl-Ala-OH

H52176.03 H52176.06
H52176.03EA 5870 CZK
H52176.03 H52176.06
Fmoc-L-3,3-diphenylalanine 95%
Fmoc-L-3,3-diphenylalanine

Purity: 95% CAS No.: 201484-50-6 Molecular Formula: C30H25NO4 Formula Weight: 463.53 Melting Point: 125-129° Beilstein Reference: 8873562 UN No.: UN3077 MDL No.: MFCD00672337

Vzorec: C₃₀H₂₅NO₄
MW: 463.533 g/mol
Bod tání: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00672337
CAS číslo: 201484-50-6
UN: 3077
ADR: 9 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR