Boc-L-beta-homophenylalanine 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Boc-L-beta-Homophe-OH, (S)-3-[(tert-Butoxycarbonyl)amino]-4-phenylbutanoic acid, Boc-β-HoPhe-OH, Boc-β-HomoPhe-OH, (S)-3-[(tert-Butoxycarbonyl)amino]-4-phenylbutyric acid, Boc-L-beta-homophenylalanine, N-(tert-Butoxycarbonyl)-L-beta-homophenylalanine
Purity: 95%
CAS No.: 51871-62-6
Molecular Formula: C15H21NO4
Formula Weight: 279.34
Melting Point: 101-107°
Beilstein Reference: 3060457
Optical Rotation: -19.0° (c=1 in dichloromethane)
MDL No.: MFCD01076271
Vzorec:
C₁₅H₂₁NO₄ Bod tání: 101…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076271 CAS číslo: 51871-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...