Tisk…

2-Allyl-D-glycine 95%

Dodavatel: Thermo Fisher Scientific

H52112.MD H52112.03
H52112.MDEA 2530 CZK
H52112.MD H52112.03
2-Allyl-D-glycine 95%
2-Allyl-D-glycine

Purity: 95% CAS No.: 54594-06-8 Molecular Formula: C5H9NO2 Formula Weight: 115.13 MDL No.: MFCD00063104

Vzorec: C₅H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00063104
CAS číslo: 54594-06-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR