Tisk…

Boc-D-Tyr(ME)-OH 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: Boc-D-Phe(4-OMe)-OH, N-Boc-4-methoxy-D-phenylalanine, N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine

H52082.03 H52082.06
H52082.03EA 1760 CZK
H52082.03 H52082.06
Boc-D-Tyr(ME)-OH 95%
Boc-D-Tyr(ME)-OH

Purity: 95% CAS No.: 68856-96-2 Molecular Formula: C15H21NO5 Formula Weight: 295.33

Vzorec: C₁₅H₂₁NO₅
Storage Temperature: Ambient
MDL Number: MFCD00065604
CAS číslo: 68856-96-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR