FMOC-L-3,4-difluorophenylalanine 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Fmoc-Phe(3,4-DiF)-OH
H52057.MD
H52057.03
H52057.06
H52057.MDEA
1970
CZK
H52057.MD
H52057.03
H52057.06
FMOC-L-3,4-difluorophenylalanine 95%
FMOC-L-3,4-difluorophenylalanine
Purity: 95%
CAS No.: 198560-43-9
Molecular Formula: C24H19F2NO4
Formula Weight: 423.40
Beilstein Reference: 8947230
MDL No.: MFCD00672550
Vzorec:
C₂₄H₁₉F₂NO₄ MW: 423.42 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00672550 CAS číslo: 198560-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...