Tisk…

(R)-3-Amino-3-phenylpropionic acid 95%

Dodavatel: Thermo Fisher Scientific
Varování

H52034.MD H52034.03 H52034.06
H52034.MDEA 2440 CZK
H52034.MD H52034.03 H52034.06
(R)-3-Amino-3-phenylpropionic acid 95%
(R)-3-Amino-3-phenylpropionic acid

Purity: 95% CAS No.: 83649-48-3 Molecular Formula: C9H11NO2 Formula Weight: 165.19 MDL No.: MFCD01076237

Vzorec: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD01076237
CAS číslo: 13921-90-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR