Tisk…

Boc-3-bromo-L-phenylalanine 95%

Dodavatel: Thermo Scientific

Synonyms: Boc-Phe(3-Br)-OH

H52024.03 H52024.06
H52024.03EA 3680 CZK
H52024.03 H52024.06
Boc-3-bromo-L-phenylalanine 95%
Boc-3-bromo-L-phenylalanine

Purity: 95% CAS No.: 82278-73-7 Molecular Formula: C14H18BrNO4 Formula Weight: 344.20 MDL No.: MFCD01317710

Vzorec: C₁₄H₁₈BrNO₄
MW: 344,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317710
CAS číslo: 82278-73-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR