Tisk…

Fmoc-Dab(Boc)-OH 96%

Dodavatel: Thermo Fisher Scientific

Synonyms: (2S)-2,4-Diaminobutanoic acid, 4-BOC, 2-FMOC protected

H51990.03 H51990.06
H51990.03EA 4290 CZK
H51990.03 H51990.06
Fmoc-Dab(Boc)-OH 96%
Fmoc-Dab(Boc)-OH

Purity: 96% CAS No.: 125238-99-5 Molecular Formula: C24H28N2O6 Formula Weight: 440.50 Melting Point: 111-113° Beilstein Reference: 6250328 Optical Rotation: -12° (c=1 in methanol) MDL No.: MFCD00151941

Vzorec: C₂₄H₂₈N₂O₆
MW: 440.496 g/mol
Bod tání: 111…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00151941
CAS číslo: 125238-99-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR