Fmoc-4-fluoro-L-phenylalanine 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-fluoro-L-phenylalanine, Fmoc-p-fluoro-Phe-OH, Fmoc-Phe(4-F)-OH, (S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3-(4-fluorophenyl)propanoic acid, N-Fmoc-4-fluoro-L-phenylalanine
H51972.03
H51972.06
H51972.03EA
4050
CZK
H51972.03
H51972.06
Fmoc-4-fluoro-L-phenylalanine 95%
Fmoc-4-fluoro-L-phenylalanine
Purity: 95%
CAS No.: 169243-86-1
Molecular Formula: C24H20FNO4
Formula Weight: 405.42
Beilstein Reference: 8287377
MDL No.: MFCD00191197
Vzorec:
C₂₄H₂₀FNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00191197 CAS číslo: 169243-86-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...