Tisk…

4-Amino-L-phenylalanine 95%

Dodavatel: Thermo Fisher Scientific

H51970.03 H51970.06
H51970.03EA 2650 CZK
H51970.03 H51970.06
4-Amino-L-phenylalanine 95%
4-Amino-L-phenylalanine

Purity: 95% CAS No.: 943-80-6 Molecular Formula: C9H12N2O2 Formula Weight: 180.20 MDL No.: MFCD00069927

Vzorec: C₉H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00069927
CAS číslo: 943-80-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR