(S)-N-Boc-4-bromophenylalanine 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Boc-p-bromo-Phe-OH, Boc-Phe(4-Br)-OH
H51969.03
H51969.06
H51969.03EA
2700
CZK
H51969.03
H51969.06
(S)-N-Boc-4-bromophenylalanine 98%
(S)-N-Boc-4-bromophenylalanine
Purity: 98%
CAS No.: 62129-39-9
Molecular Formula: C14H18BrNO4
Formula Weight: 344.20
Melting Point: 118-120°
MDL No.: MFCD00237571
Vzorec:
C₁₄H₁₈BrNO₄ MW: 344.2 g/mol Bod tání: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00237571 CAS číslo: 62129-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...