Tisk…

Anthracene-9,10-diboronic acid bis(pinacol) ester 95%

Dodavatel: Thermo Fisher Scientific

H51921.03 H51921.06
H51921.03EA 3030 CZK
H51921.03 H51921.06
Anthracene-9,10-diboronic acid bis(pinacol) ester 95%
Anthracene-9,10-diboronic acid bis(pinacol) ester

Purity: 95% CAS No.: 863992-56-7 Molecular Formula: C26H32B2O4 Formula Weight: 430.16 MDL No.: MFCD16294538

Vzorec: C₂₆H₃₂B₂O₄
Storage Temperature: Ambient
MDL Number: MFCD16294538
CAS číslo: 863992-56-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR