Tisk…

4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: tert-Butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine-1-carboxylate

H51743.03 H51743.06
H51743.03EA 2480 CZK
H51743.03 H51743.06
4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%
4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester

Purity: 95% CAS No.: 936694-19-8 Molecular Formula: C22H35BN2O4 Formula Weight: 402.34 Beilstein Reference: 18669672 MDL No.: MFCD16294502

Vzorec: C₂₂H₃₅BN₂O₄
MW: 402.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294502
CAS číslo: 936694-19-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR