(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
(S)-tert-Butyl pyrrolidin-3-ylcarbamate, (3S)-(-)-3-(Boc-amino)pyrrolidine
H51729.06
H51729.14
H51729.06EA
4530
CZK
H51729.06
H51729.14
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine
Purity: 98%
CAS No.: 122536-76-9
Molecular Formula: C9H18N2O2
Formula Weight: 186.25
Melting Point: 50-54°
Beilstein Reference: 5377813
Optical Rotation: -23° (c=1 in methanol)
UN No.: UN3259
MDL No.: MFCD00143194
Vzorec:
C₉H₁₈N₂O₂ MW: 186,26 g/mol Bod tání: 50…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143194 CAS číslo: 122536-76-9 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...