Thiopicolinamide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H51710.06
H51710.14
H51710.06EA
1270
CZK
H51710.06
H51710.14
Thiopicolinamide ≥97%
Thiopicolinamide
Purity: 97+%
CAS No.: 5346-38-3
Molecular Formula: C6H6N2S
Formula Weight: 138.19
Melting Point: 120-125°
MDL No.: MFCD00087576
Vzorec:
C₆H₆N₂S Bod tání: 120…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00087576 CAS číslo: 5346-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...