Tisk…

1,4-Benzenediboronic acid bis(pinacol)ester 97%

Dodavatel: Thermo Scientific

Synonyms: 1,4-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 1,4-Bis(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 1,4-Phenylenediboronic acid, pinacol ester

H51707.03 H51707.06
H51707.03EA 2750 CZK
H51707.03 H51707.06
1,4-Benzenediboronic acid bis(pinacol)ester 97%
1,4-Benzenediboronic acid bis(pinacol)ester

Purity: 97% CAS No.: 99770-93-1 Molecular Formula: C18H28B2O4 Formula Weight: 330.03 MDL No.: MFCD08276852

Vzorec: C₁₈H₂₈B₂O₄
MW: 330,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276852
CAS číslo: 99770-93-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR