Tisk…

Ethyl-5-aminovalerate hydrochloride 96%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl 5-aminopentanoate hydrochloride

H51682.MD H51682.03
H51682.MDEA 2410 CZK
H51682.MD H51682.03
Ethyl-5-aminovalerate hydrochloride 96%
Ethyl-5-aminovalerate hydrochloride

Purity: 96% CAS No.: 29840-57-1 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 MDL No.: MFCD12546205

Vzorec: C₇H₁₆ClNO₂
MW: 181,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546205
CAS číslo: 29840-57-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR