N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
Dodavatel: Thermo Scientific
H51136.03
H51136.06
H51136.14
H51136.03EA
1130
CZK
H51136.03
H51136.06
H51136.14
N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
N-(tert-Butoxycarbonyl)-L-tert-leucine
Purity: 98%
CAS No.: 62965-35-9
Molecular Formula: C11H21NO4
Formula Weight: 231.29
Melting Point: 118-121°
MDL No.: MFCD00065574
Vzorec:
C₁₁H₂₁NO₄ Bod tání: 118…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065574 CAS číslo: 62965-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...