tert-Butyl methyl(2-oxoethyl)carbamate 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
tert-Butyl methyll(2-oxoethyl)carbamate, Methyl(2-oxoethyl)carbamic acid tert-butyl ester, N-Boc-(methylamino)acetaldehyde, N-tert-Butoxycarbonyl-(methylamino)acetaldehyde
H50870.03
H50870.MD
H50870.03EA
6040
CZK
H50870.03
H50870.MD
tert-Butyl methyl(2-oxoethyl)carbamate 97%
tert-Butyl methyl(2-oxoethyl)carbamate
Purity: 98%
CAS No.: 123387-72-4
Molecular Formula: C8H15NO3
Formula Weight: 173.21
Storage Temperature: 2°C to 8°C
MDL No.: MFCD08064267
Vzorec:
C₈H₁₅NO₃ MW: 173.21 g/mol |
MDL Number:
MFCD08064267 CAS číslo: 123387-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...