3-(2-Methylphenoxy)pyrrolidine 95%
Dodavatel: Thermo Scientific
|
Varování
|
H50784.03
H50784.MD
H50784.03EA
4530
CZK
H50784.03
H50784.MD
3-(2-Methylphenoxy)pyrrolidine 95%
3-(2-Methylphenoxy)pyrrolidine
Purity: 95%
CAS No.: 46196-54-7
Molecular Formula: C11H15NO
Formula Weight: 177.25
MDL No.: MFCD08060984
Vzorec:
C₁₁H₁₅NO Storage Temperature: Ambient |
MDL Number:
MFCD08060984 CAS číslo: 46196-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...